Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160127.1 | NP_001153599.1 | 8 - 9 | 1220 | 64 | |
NM_009762.2 | NP_033892.2 | 7 - 8 | 1181 | 64 | ||
GenBank mRNA | AK076266.1 | - | 7 - 8 | 1146 | 64 | |
AK142153.1 | - | 7 - 8 | 1149 | 64 | ||
AK142252.1 | - | 8 - 9 | 1198 | 64 | ||
BC076601.1 | - | 8 - 9 | 1198 | 30609878 | 64 | |
U76373.2 | - | 8 - 9 | 1192 | 64 | ||
U76374.2 | - | 7 - 8 | 1153 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |