Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010937.2 | NP_035067.2 | 6 - 7 | 789 | 131 | |
GenBank mRNA | AK010412.1 | - | 6 - 7 | 750 | 131 | |
AK011704.1 | - | 6 - 7 | 736 | 131 | ||
AK150357.1 | - | 6 - 7 | 728 | 131 | ||
AK165960.1 | - | 6 - 7 | 733 | 131 | ||
BC014704.1 | - | 6 - 7 | 717 | 131 | ||
BC051443.1 | - | 6 - 7 | 712 | 30061371 | 131 | |
BC058755.1 | - | 6 - 7 | 721 | 6475312 | 131 | |
X13664.1 | - | 6 - 7 | 832 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |