Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011059.2 | NP_035189.1 | 9 - 10 | 1177 | 63 | |
GenBank mRNA | AB013848.1 | - | 9 - 10 | 1177 | 63 | |
AK037182.1 | - | 9 - 10 | 1178 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |