Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011782.2 | NP_035912.2 | 4 - 5 | 2542 | 103 | |
GenBank mRNA | AF140673.1 | - | 4 - 5 | 1701 | 103 | |
AK044746.1 | - | 4 - 5 | 764 | 103 | ||
AK082475.1 | - | 4 - 5 | 764 | 103 | ||
BC138619.1 | - | 4 - 5 | 1726 | 8861639 | 103 | |
BC138620.1 | - | 4 - 5 | 1726 | 8861640 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |