Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893307 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893306 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893305 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893308 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2023523 |
Y14695.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2023523 |
NM_011786.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
504386 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408288 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689419 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
307379 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379276 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821134 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770211 |
XM_001478763.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770211 |
XM_917116.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
830900 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702398 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291973 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
646098 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898104 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
199425 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484267 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
953900 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5316 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
73779 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
166731 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160391 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160096 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2023523 |
Y14695.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2023523 |
NM_011786.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P136143 |
Y14695.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P136143 |
NM_011786.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893235 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893244 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893241 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893257 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893251 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893238 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893255 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893246 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893262 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893231 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893259 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893223 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893228 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893230 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893249 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893240 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893229 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893222 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893225 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893248 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893258 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893253 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27893233 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07384623 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07384583 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160096 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160391 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02354982 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P136143 |
Y14695.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P136143 |
NM_011786.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130829 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160391 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160096 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112599 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112608 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112607 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112601 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112595 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09112605 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893307 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893305 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893306 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27893308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893398 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893312 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27893266 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09215806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893227 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09317022 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893249 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893240 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09316998 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09112608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893251 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09734296 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09661055 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893276 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893330 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893288 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09420138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893277 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09154346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893333 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09317009 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893225 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09112599 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893297 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893359 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09112596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893231 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893271 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893295 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893274 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893335 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893306 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09112594 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893260 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893255 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893253 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893358 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893286 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893228 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893266 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893223 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09661054 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893279 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09316991 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893300 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09656537 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893238 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893282 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893303 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09608724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09317025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893222 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893299 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893293 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893292 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09132456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09420132 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09608728 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27893229 |
- |
- |