Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013628.2 | NP_038656.1 | 9 - 10 | 1271 | 67 | |
GenBank mRNA | BC108982.1 | - | 9 - 10 | 1249 | 40047652 | 67 |
BC108983.1 | - | 9 - 10 | 1249 | 40047655 | 67 | |
M55668.1 | - | 4 - 5 | 557 | 67 | ||
M58589.1 | - | 9 - 10 | 1271 | 67 | ||
M69196.1 | - | 9 - 10 | 1377 | 67 | ||
X57088.1 | - | 9 - 10 | 1383 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |