Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018814.3 | NP_061284.2 | 14 - 15 | 3699 | 88 | |
GenBank mRNA | AF096286.1 | - | 8 - 9 | 1529 | 88 | |
AK129221.1 | - | 9 - 10 | 1579 | 88 | ||
BC057333.1 | - | 7 - 8 | 1442 | 88 | ||
BC098370.1 | - | 9 - 10 | 1289 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |