Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY082613.1 | - | 1 - 2 | 95 | 99 | |
U38981.1 | - | 208 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |