Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019811.3 | NP_062785.2 | 7 - 8 | 896 | 65 | |
GenBank mRNA | AF216873.1 | - | 7 - 8 | 846 | 65 | |
AK028586.1 | - | 7 - 8 | 874 | 65 | ||
AK029014.1 | - | 7 - 8 | 864 | 65 | ||
AK029262.1 | - | 882 | 65 | |||
AK053877.1 | - | 7 - 8 | 870 | 65 | ||
AK093831.1 | - | 7 - 8 | 873 | 65 | ||
BC040790.1 | - | 7 - 8 | 846 | 65 | ||
BC051432.1 | - | 7 - 8 | 896 | 6515568 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |