Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019933.2 | NP_064317.2 | 5 - 6 | 558 | 103 | |
GenBank mRNA | AF106702.1 | - | 4 - 5 | 386 | 103 | |
AK046683.1 | - | 5 - 6 | 833 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus Probe spans exons |