Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039509.1 | NP_001034598.1 | 5 - 6 | 559 | 128 | |
NM_019999.2 | NP_064383.1 | 4 - 5 | 468 | 128 | ||
GenBank mRNA | AB041609.1 | - | 4 - 5 | 469 | 128 | |
AK041238.1 | - | 4 - 5 | 402 | 128 | ||
AK173125.1 | - | 5 - 6 | 533 | 128 | ||
BC058945.1 | - | 6 - 7 | 655 | 6812127 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |