Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020561.2 | NP_065586.3 | 3 - 4 | 599 | 70 | |
GenBank mRNA | AK002455.1 | - | 3 - 4 | 538 | 70 | |
AK151683.1 | - | 3 - 4 | 485 | 70 | ||
AK152282.1 | - | 3 - 4 | 500 | 70 | ||
AK185060.1 | - | 2 - 3 | 345 | 70 | ||
AK194580.1 | - | 2 - 3 | 345 | 70 | ||
AK205531.1 | - | 2 - 3 | 348 | 70 | ||
BC139288.1 | - | 3 - 4 | 491 | 8862310 | 70 | |
BC139289.1 | - | 3 - 4 | 491 | 8862311 | 70 | |
Y08135.1 | - | 3 - 4 | 553 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |