Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021320.3 | NP_067295.2 | 1 - 2 | 461 | 68 | |
GenBank mRNA | AF268066.1 | - | 1 - 2 | 56 | 68 | |
AF281278.1 | - | 1 - 2 | 309 | 68 | ||
BB609851.1 | - | 1 - 2 | 332 | 68 | ||
BC119182.1 | - | 1 - 2 | 143 | 8733931 | 68 | |
BC119184.1 | - | 1 - 2 | 143 | 8733933 | 68 | |
CJ068258.1 | - | 1 - 2 | 461 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |