Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037762.1 | NP_001032851.1 | 2 - 3 | 290 | 61 | |
NM_025428.2 | NP_079704.2 | 2 - 3 | 332 | 61 | ||
GenBank mRNA | AK004475.1 | - | 2 - 3 | 233 | 61 | |
AK013074.1 | - | 2 - 3 | 320 | 61 | ||
AK087676.1 | - | 2 - 3 | 304 | 61 | ||
BC021432.1 | - | 2 - 3 | 272 | 5030263 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |