Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025453.3 | NP_079729.1 | 1 - 2 | 225 | 75 | |
GenBank mRNA | AK007532.1 | - | 1 - 2 | 226 | 75 | |
AK017154.1 | - | 1 - 2 | 227 | 75 | ||
AK017195.1 | - | 1 - 2 | 226 | 75 | ||
AK017209.1 | - | 1 - 2 | 234 | 75 | ||
BC117736.2 | - | 1 - 2 | 217 | 40089849 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |