Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025548.3 | NP_079824.2 | 1 - 2 | 452 | 104 | |
GenBank mRNA | AK002316.1 | - | 1 - 2 | 193 | 104 | |
AK003655.2 | - | 1 - 2 | 198 | 104 | ||
AK010438.1 | - | 1 - 2 | 209 | 104 | ||
BC010684.1 | - | 1 - 2 | 185 | 2648112 | 104 | |
BQ445322.1 | - | 1 - 2 | 452 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |