Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 7 - 8 | 687 | 77 | |
GenBank mRNA | AK009346.1 | - | 7 - 8 | 660 | 77 | |
AK009504.1 | - | 7 - 8 | 662 | 77 | ||
AK009619.1 | - | 7 - 8 | 660 | 77 | ||
AK009754.1 | - | 3 - 4 | 263 | 77 | ||
AK009794.1 | - | 7 - 8 | 658 | 77 | ||
AK009871.1 | - | 7 - 8 | 662 | 77 | ||
AK010130.1 | - | 7 - 8 | 644 | 77 | ||
AK010154.1 | - | 7 - 8 | 658 | 77 | ||
AK010157.1 | - | 7 - 8 | 688 | 77 | ||
AK010215.1 | - | 7 - 8 | 658 | 77 | ||
AK028156.1 | - | 7 - 8 | 658 | 77 | ||
AK028158.1 | - | 7 - 8 | 661 | 77 | ||
AK075874.1 | - | 7 - 8 | 658 | 77 | ||
AK160516.1 | - | 7 - 8 | 658 | 77 | ||
BC116681.1 | - | 7 - 8 | 642 | 40106489 | 77 | |
BC116727.1 | - | 7 - 8 | 40106498 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |