Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 45 - 46 | 6829 | 76 | |
GenBank mRNA | AK014886.1 | - | 6 - 7 | 888 | 76 | |
AK014938.1 | - | 6 - 7 | 901 | 76 | ||
AY792512.1 | - | 45 - 46 | 6772 | 76 | ||
BC034074.1 | - | 11 - 12 | 1712 | 76 | ||
BC035949.1 | - | 1 - 2 | 173 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |