Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
AB055902.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
AK048146.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
AK052640.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
AK136293.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
AK220433.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
BC056368.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
BC058082.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
BC092280.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572627 |
NM_172124.2 |
refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169898 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055927 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055941 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055970 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055947 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055962 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055971 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055930 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055972 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055976 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055940 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055967 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055965 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055958 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055916 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055920 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055961 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055954 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055917 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055925 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055919 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055973 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055943 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055924 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055938 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055968 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055946 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055932 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055953 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055949 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055951 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055957 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055959 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055934 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055974 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055952 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32055922 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169898 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
BC125314.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
BC125316.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
NM_025741.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
AK016362.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P163924 |
AK016450.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117266 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169898 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14455248 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14383361 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14455266 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14455257 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14455275 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14383360 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14383363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32056005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32055938 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14455265 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055954 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14461610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055990 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055949 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14161363 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055961 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055985 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055952 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055970 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055951 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055943 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055917 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055945 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055915 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14455276 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055931 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055916 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32056006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14239005 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32056002 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14455258 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055968 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055934 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14287794 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055938 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055941 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055986 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055979 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14383360 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055947 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14374156 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32056005 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055972 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055946 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055920 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055967 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055935 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055982 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055953 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055932 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14461613 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32056000 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055927 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055957 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055925 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14161356 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32055994 |
- |
- |