Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2012221 |
BC054839.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2012221 |
BE986475.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2012221 |
BC049088.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2012221 |
BC052383.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2012221 |
AK172960.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
445707 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563329 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593080 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330600 |
BC119113.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330600 |
BC119115.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330600 |
NM_146985.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
812227 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
321364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
714218 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
775001 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
612257 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
684691 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81127 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
164447 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P159155 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2012221 |
BC054839.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2012221 |
BE986475.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2012221 |
BC049088.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2012221 |
BC052383.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2012221 |
AK172960.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673765 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673768 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673775 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673769 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673777 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673791 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673795 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673798 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673803 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673789 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673780 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673779 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673751 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673783 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673760 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673773 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673753 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673785 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673767 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673772 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673786 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673754 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673782 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673800 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673750 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673774 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673776 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673793 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673752 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673761 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32673801 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09426593 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09426620 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P159155 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P356512 |
BC054839.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356512 |
BE986475.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356512 |
BC049088.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356512 |
BC052383.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P356512 |
AK172960.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106295 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P159155 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092220 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092236 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092231 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092218 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092237 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092233 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15092221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32673782 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673801 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673663 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673659 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14610891 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673785 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673829 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673835 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673725 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673767 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673665 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673820 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673658 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15092220 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673666 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673744 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840248 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673798 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673777 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673821 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673811 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14592721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673718 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673728 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673789 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673761 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673758 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673831 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673751 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673786 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673802 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673749 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673824 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673775 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673765 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673808 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673822 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673752 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14610908 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673739 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673819 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673669 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840229 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673661 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673667 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14982251 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673772 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673809 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14840231 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14622522 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673671 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673827 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673746 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673769 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673803 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14610890 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15092221 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673780 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673660 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673782 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673768 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14726529 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14618373 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673754 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673730 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673742 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673773 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32673719 |
- |
- |