Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007636.2 | NP_031662.2 | 12 - 13 | 1339 | 81 | |
GenBank mRNA | AK012295.1 | - | 12 - 13 | 1299 | 81 | |
AK075581.1 | - | 12 - 13 | 1312 | 81 | ||
AK088145.1 | - | 12 - 13 | 1301 | 81 | ||
AK145406.1 | - | 12 - 13 | 1340 | 81 | ||
AK166891.1 | - | 12 - 13 | 1312 | 81 | ||
BC007470.1 | - | 12 - 13 | 1303 | 3601569 | 81 | |
BC026918.1 | - | 12 - 13 | 1305 | 4500896 | 81 | |
Z31553.1 | - | 12 - 13 | 1290 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |