Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007642.4 | NP_031668.3 | 1 - 2 | 132 | 68 | |
GenBank mRNA | AK030812.1 | - | 1 - 2 | 54 | 68 | |
AK041322.1 | - | 1 - 2 | 119 | 68 | ||
AK088021.1 | - | 1 - 2 | 65 | 68 | ||
BC064058.1 | - | 1 - 2 | 46 | 3418931 | 68 | |
M34563.1 | - | 1 - 2 | 102 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Mouse Tumor Metastasis Panel Mouse Immune Response Inventoried Mouse_Immune_Response Probe spans exons |