Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252525.1 | NP_001239454.1 | 7 - 8 | 1184 | 69 | |
NM_007755.5 | NP_031781.1 | 7 - 8 | 1181 | 69 | ||
GenBank mRNA | AK136088.1 | - | 7 - 8 | 1185 | 69 | |
BC125476.1 | - | 7 - 8 | 1101 | 40129991 | 69 | |
Y08260.1 | - | 7 - 8 | 1100 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |