Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007810.3 | NP_031836.1 | 4 - 5 | 668 | 90 | |
GenBank mRNA | AK143323.1 | - | 4 - 5 | 748 | 90 | |
BC103670.1 | - | 4 - 5 | 651 | 40047093 | 90 | |
BC104489.1 | - | 4 - 5 | 651 | 40047095 | 90 | |
D00659.1 | - | 4 - 5 | 673 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |