Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008012.1 | NP_032038.1 | 7 - 8 | 804 | 58 | |
GenBank mRNA | U04204.1 | - | 7 - 8 | 804 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |