Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200046067.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046067.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046067.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046072.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046072.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046072.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046073.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046073.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046073.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046068.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046068.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046068.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046071.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046071.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046071.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046075.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046075.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046075.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046069.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046069.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046069.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046074.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046074.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046074.mm.1 |
TC0200005408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046070.mm.1 |
NM_008093.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046070.mm.1 |
NM_008093.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046070.mm.1 |
TC0200005408.mm.1 |
MTA TC |