Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008608.3 | NP_032634.3 | 1 - 2 | 376 | 61 | |
GenBank mRNA | AK040968.1 | - | 347 | 61 | ||
AK088476.1 | - | 1 - 2 | 376 | 61 | ||
AK134340.1 | - | 1 - 2 | 307 | 61 | ||
AK138611.1 | - | 1 - 2 | 317 | 61 | ||
AK149636.1 | - | 1 - 2 | 305 | 61 | ||
AK149907.1 | - | 1 - 2 | 356 | 61 | ||
AK149988.1 | - | 1 - 2 | 276 | 61 | ||
AK155117.1 | - | 1 - 2 | 378 | 61 | ||
AK163601.1 | - | 1 - 2 | 368 | 61 | ||
AK165014.1 | - | 1 - 2 | 377 | 61 | ||
AK198018.1 | - | 1 - 2 | 90 | 61 | ||
BC076638.1 | - | 1 - 2 | 368 | 30658833 | 61 | |
DQ249870.1 | - | 1 - 2 | 114 | 61 | ||
U54984.1 | - | 1 - 2 | 360 | 61 | ||
X83536.2 | - | 1 - 2 | 286 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus Mouse Tumor Metastasis Panel 1700 Inventoried Probe spans exons |