Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009014.3 | NP_033040.2 | 9 - 10 | 1030 | 68 | |
GenBank mRNA | AK032913.1 | - | 9 - 10 | 1031 | 68 | |
U92068.1 | - | 8 - 9 | 972 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |