Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009043.1 | NP_033069.1 | 3 - 4 | 272 | 96 | |
GenBank mRNA | AK008196.1 | - | 3 - 4 | 271 | 96 | |
BC098503.1 | - | 2 - 3 | 255 | 6431377 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |