Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009118.2 | NP_033144.1 | 2 - 3 | 438 | 79 | |
GenBank mRNA | AK044214.1 | - | 1 - 2 | 119 | 79 | |
AK053278.1 | - | 2 - 3 | 197 | 79 | ||
AK143098.1 | - | 2 - 3 | 197 | 79 | ||
AK149299.1 | - | 2 - 3 | 447 | 79 | ||
AY651760.1 | - | 2 - 3 | 226 | 79 | ||
BC016498.1 | - | 2 - 3 | 231 | 4527551 | 79 | |
M24086.1 | - | 2 - 3 | 225 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |