Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163630.1 | NP_001157102.1 | 5 - 6 | 965 | 72 | |
NM_001163631.1 | NP_001157103.1 | 5 - 6 | 833 | 72 | ||
NM_001163632.1 | NP_001157104.1 | 5 - 6 | 806 | 72 | ||
NM_009122.2 | NP_033148.2 | 5 - 6 | 856 | 72 | ||
GenBank mRNA | AK037740.1 | - | 5 - 6 | 856 | 72 | |
AK088563.1 | - | 5 - 6 | 832 | 72 | ||
AK134040.1 | - | 5 - 6 | 836 | 72 | ||
AK158518.1 | - | 5 - 6 | 966 | 72 | ||
BC011132.1 | - | 5 - 6 | 716 | 4164993 | 72 | |
CT010371.1 | - | 4 - 5 | 644 | 72 | ||
U05252.1 | - | 5 - 6 | 842 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |