Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169679 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169681 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169672 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169675 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169677 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169680 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169673 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169678 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2024530 |
BC058400.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2024530 |
NM_009174.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
109145 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851231 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
872219 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
181156 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685993 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
133708 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
701371 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
854180 |
AK005130.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
854180 |
AK154249.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
854180 |
NM_025895.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
629236 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933743 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
784819 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
32342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178950 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P180340 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P109666 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2024530 |
BC058400.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2024530 |
NM_009174.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P183803 |
Z19581.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P183803 |
AK149316.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P183803 |
BC058400.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P183803 |
NM_009174.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169686 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169684 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169700 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169678 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169675 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169714 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169661 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169679 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169664 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169694 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169705 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169663 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169682 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169697 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169659 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169717 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169672 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169711 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169665 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169668 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169689 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169692 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169699 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22169670 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P180340 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P109666 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P183803 |
Z19581.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P183803 |
AK149316.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P183803 |
BC058400.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P183803 |
NM_009174.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103460 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P109666 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P180340 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03038477 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02788103 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02788102 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02385160 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169681 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169680 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169679 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169677 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169675 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169678 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169672 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22169673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22169678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03196084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169633 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169714 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02407150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908560 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169722 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169596 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169646 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169642 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169688 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03145678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169659 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169715 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169650 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169583 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169663 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02407151 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908573 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169585 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02431962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02862359 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908572 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02565896 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03145681 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02431951 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169661 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02741535 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169694 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169664 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169717 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169686 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908567 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169726 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169588 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02803523 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02433224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169656 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169576 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169586 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03024774 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169595 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03038477 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02918571 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02918578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03196085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02661362 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02333711 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169724 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02333719 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02385160 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169721 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02433225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908564 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169654 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02817196 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22169644 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02908575 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02803522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02788102 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02624867 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02624868 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02788103 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02918572 |
- |
- |