Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253804.1 | NP_001240733.1 | 2 - 3 | 295 | 76 | |
NM_009195.3 | NP_033221.1 | 2 - 3 | 289 | 76 | ||
NR_045594.1 | 2 - 3 | 289 | 76 | |||
GenBank mRNA | AF047339.1 | - | 2 - 3 | 213 | 76 | |
AF121118.1 | - | 2 - 3 | 280 | 76 | ||
AK149969.1 | - | 2 - 3 | 278 | 76 | ||
AK151717.1 | - | 2 - 3 | 250 | 76 | ||
AK152762.1 | - | 2 - 3 | 267 | 76 | ||
AK153245.1 | - | 2 - 3 | 267 | 76 | ||
AK159274.1 | - | 2 - 3 | 277 | 76 | ||
AK159331.1 | - | 2 - 3 | 272 | 76 | ||
AK159483.1 | - | 2 - 3 | 245 | 76 | ||
AK159537.1 | - | 2 - 3 | 272 | 76 | ||
AK160034.1 | - | 2 - 3 | 238 | 76 | ||
AK160072.1 | - | 2 - 3 | 272 | 76 | ||
AK164479.1 | - | 2 - 3 | 254 | 76 | ||
AK165874.1 | - | 2 - 3 | 267 | 76 | ||
AK166304.1 | - | 2 - 3 | 248 | 76 | ||
AK167371.1 | - | 2 - 3 | 286 | 76 | ||
AK167444.1 | - | 2 - 3 | 286 | 76 | ||
AK169044.1 | - | 2 - 3 | 244 | 76 | ||
BC066872.1 | - | 2 - 3 | 219 | 30436525 | 76 | |
BY740705.1 | - | 2 - 3 | 290 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |