Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164639.1 | NP_001158111.1 | 16 - 17 | 3523 | 61 | |
NM_009289.3 | NP_033315.2 | 17 - 18 | 3616 | 61 | ||
GenBank mRNA | AF039574.1 | - | 17 - 18 | 3621 | 61 | |
AF112855.1 | - | 16 - 17 | 3412 | 61 | ||
AK037798.1 | - | 8 - 9 | 1497 | 61 | ||
AK122218.1 | - | 16 - 17 | 3864 | 61 | ||
BC131674.1 | - | 16 - 17 | 3424 | 40110915 | 61 | |
BC131675.1 | - | 17 - 18 | 3517 | 40110921 | 61 | |
CO431234.1 | - | 3 - 4 | 404 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |