Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009302.3 | NP_033328.3 | 5 - 6 | 898 | 149 | |
GenBank mRNA | AF053974.1 | - | 5 - 6 | 790 | 149 | |
AK019882.1 | - | 5 - 6 | 842 | 149 | ||
AK146773.1 | - | 5 - 6 | 890 | 149 | ||
AK172990.1 | - | 5 - 6 | 835 | 149 | ||
BC065136.1 | - | 5 - 6 | 840 | 30546814 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |