Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110228.1 | NP_001103698.1 | 9 - 10 | 1049 | 85 | |
NM_001110229.1 | NP_001103699.1 | 10 - 11 | 1136 | 85 | ||
NM_001110230.1 | NP_001103700.1 | 10 - 11 | 1136 | 85 | ||
NM_001110231.1 | NP_001103701.1 | 12 - 13 | 1111 | 85 | ||
NM_001110232.1 | NP_001103702.1 | 11 - 12 | 1122 | 85 | ||
NM_001160292.1 | NP_001153764.1 | 10 - 11 | 1136 | 85 | ||
NM_001160293.1 | NP_001153765.1 | 9 - 10 | 1049 | 85 | ||
NM_010160.2 | NP_034290.2 | 9 - 10 | 1049 | 85 | ||
GenBank mRNA | AF090696.1 | - | 11 - 12 | 1122 | 85 | |
AF090697.1 | - | 9 - 10 | 1022 | 85 | ||
AK135166.1 | - | 9 - 10 | 1001 | 85 | ||
AK137292.1 | - | 10 - 11 | 1077 | 85 | ||
AK151818.1 | - | 8 - 9 | 907 | 85 | ||
AK152628.1 | - | 8 - 9 | 907 | 85 | ||
AK153267.1 | - | 8 - 9 | 907 | 85 | ||
AK159996.1 | - | 10 - 11 | 1141 | 85 | ||
AK160861.1 | - | 12 - 13 | 1112 | 85 | ||
AK161349.1 | - | 2 - 3 | 445 | 85 | ||
AK164914.1 | - | 10 - 11 | 1077 | 85 | ||
BC026856.1 | - | 9 - 10 | 1000 | 4503295 | 85 | |
Y18298.1 | - | 8 - 9 | 915 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |