Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161798.1 | NP_001155270.1 | 10 - 11 | 1772 | 84 | |
NM_010840.3 | NP_034970.2 | 10 - 11 | 1695 | 84 | ||
NR_027809.1 | 10 - 11 | 1772 | 84 | |||
GenBank mRNA | AK030192.1 | - | 10 - 11 | 1690 | 84 | |
AK043144.1 | - | 2 - 3 | 121 | 84 | ||
AK149795.1 | - | 10 - 11 | 1757 | 84 | ||
AK150159.1 | - | 10 - 11 | 1773 | 84 | ||
BC051017.1 | - | 10 - 11 | 1674 | 6308248 | 84 | |
BC052466.1 | - | 10 - 11 | 1739 | 6834886 | 84 | |
BC100550.1 | - | 2 - 3 | 163 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |