Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271407.1 | NP_001258336.1 | 12 - 13 | 1300 | 72 | |
NM_001271408.1 | NP_001258337.1 | 12 - 13 | 1184 | 72 | ||
NM_001271409.1 | NP_001258338.1 | 1 - 2 | 165 | 72 | ||
NM_011366.3 | NP_035496.1 | 12 - 13 | 1187 | 72 | ||
GenBank mRNA | AB190911.1 | - | 12 - 13 | 1301 | 72 | |
AF064806.1 | - | 12 - 13 | 1153 | 72 | ||
AK029703.1 | - | 14 - 15 | 1298 | 72 | ||
AK165689.1 | - | 12 - 13 | 1194 | 72 | ||
BC030933.1 | - | 11 - 12 | 1001 | 4920799 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |