Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011797.2 | NP_035927.1 | 1 - 2 | 322 | 69 | |
GenBank mRNA | AB005450.1 | - | 1 - 2 | 322 | 69 | |
AK157764.1 | - | 1 - 2 | 322 | 69 | ||
BC046995.1 | - | 1 - 2 | 304 | 6491779 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |