Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082414.2 | NP_001075883.2 | 7 - 8 | 1689 | 102 | |
GenBank mRNA | BC028254.1 | - | 7 - 8 | 1237 | 102 | |
BC031117.1 | - | 7 - 8 | 800 | 5064197 | 102 | |
D89677.2 | - | 13 - 14 | 2175 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |