Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033713.2 | NP_001028885.1 | 3 - 4 | 392 | 89 | |
NM_001291191.1 | NP_001278120.1 | 3 - 4 | 412 | 89 | ||
NM_001291192.1 | NP_001278121.1 | 3 - 4 | 412 | 89 | ||
NM_001291195.1 | NP_001278124.1 | 3 - 4 | 392 | 89 | ||
NM_001291196.1 | NP_001278125.1 | 3 - 4 | 392 | 89 | ||
GenBank mRNA | AK052385.1 | - | 3 - 4 | 393 | 89 | |
AK132678.1 | - | 3 - 4 | 274 | 89 | ||
AK153751.1 | - | 3 - 4 | 413 | 89 | ||
BC019116.1 | - | 3 - 4 | 378 | 89 | ||
BC061128.1 | - | 3 - 4 | 363 | 6593862 | 89 | |
BC096598.1 | - | 3 - 4 | 354 | 4979487 | 89 | |
U30823.1 | - | 1 - 2 | 141 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |