Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290717.1 | NP_001277646.1 | 12 - 13 | 1510 | 93 | |
NM_001290718.1 | NP_001277647.1 | 12 - 13 | 1483 | 93 | ||
NM_001290719.1 | NP_001277648.1 | 12 - 13 | 1510 | 93 | ||
NM_013757.2 | NP_038785.1 | 12 - 13 | 1510 | 93 | ||
GenBank mRNA | AB025258.1 | - | 12 - 13 | 1481 | 93 | |
AB025259.1 | - | 12 - 13 | 1481 | 93 | ||
AB120510.1 | - | 11 - 12 | 1296 | 93 | ||
AK030401.1 | - | 12 - 13 | 1357 | 93 | ||
AK161582.1 | - | 12 - 13 | 1476 | 93 | ||
BC026819.1 | - | 12 - 13 | 1510 | 4488012 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |