Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025391.2 | NP_001020562.1 | 7 - 8 | 1117 | 108 | |
NM_015752.3 | NP_056567.2 | 7 - 8 | 1120 | 108 | ||
GenBank mRNA | AF134893.1 | - | 7 - 8 | 1060 | 108 | |
AJ131692.1 | - | 7 - 8 | 1118 | 108 | ||
AJ308625.1 | - | 7 - 8 | 796 | 108 | ||
AJ308626.1 | - | 7 - 8 | 1051 | 108 | ||
AK015885.1 | - | 6 - 7 | 878 | 108 | ||
AK016976.1 | - | 7 - 8 | 1098 | 108 | ||
AK047603.1 | - | 7 - 8 | 990 | 108 | ||
AK156410.1 | - | 7 - 8 | 1082 | 108 | ||
BC048168.1 | - | 7 - 8 | 1090 | 6821477 | 108 | |
BC056997.1 | - | 7 - 8 | 1090 | 6821477 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |