Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016695.3 | NP_057904.1 | 4 - 5 | 428 | 67 | |
GenBank mRNA | AF162685.1 | - | 4 - 5 | 415 | 67 | |
AK141422.1 | - | 3 - 4 | 426 | 67 | ||
AK161577.1 | - | 4 - 5 | 421 | 67 | ||
AK172253.1 | - | 4 - 5 | 413 | 67 | ||
BC053026.1 | - | 4 - 5 | 428 | 5709543 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |