Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252347.1 | NP_001239276.1 | 5 - 6 | 815 | 73 | |
NM_016759.4 | NP_058039.1 | 5 - 6 | 815 | 73 | ||
GenBank mRNA | AK002856.1 | - | 1 - 2 | 113 | 73 | |
BC022745.1 | - | 3 - 4 | 540 | 73 | ||
BC046319.1 | - | 5 - 6 | 715 | 6391042 | 73 | |
U73941.1 | - | 5 - 6 | 542 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |