Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016870.3 | NP_058566.3 | 8 - 9 | 1400 | 77 | |
NM_212441.2 | NP_997606.2 | 7 - 8 | 1295 | 77 | ||
NM_212442.2 | NP_997607.2 | 7 - 8 | 1198 | 77 | ||
GenBank mRNA | AB022340.2 | - | 7 - 8 | 1186 | 77 | |
AF068246.1 | - | 7 - 8 | 1185 | 77 | ||
AK033461.1 | - | 2 - 3 | 266 | 77 | ||
AK077247.1 | - | 7 - 8 | 1261 | 77 | ||
AK143946.1 | - | 1198 | 77 | |||
AK165516.1 | - | 7 - 8 | 1172 | 77 | ||
AY064696.1 | - | 7 - 8 | 1271 | 77 | ||
BC015248.1 | - | 8 - 9 | 1360 | 4239288 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |