Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016911.4 | NP_058607.1 | 4 - 5 | 643 | 86 | |
GenBank mRNA | AB028049.1 | - | 4 - 5 | 570 | 86 | |
AB028050.1 | - | 2 - 3 | 318 | 86 | ||
AK034696.1 | - | 4 - 5 | 645 | 86 | ||
AK159662.1 | - | 3 - 4 | 631 | 86 | ||
BC022572.1 | - | 4 - 5 | 599 | 3993025 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |