Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017474.2 | NP_059502.1 | 8 - 9 | 1389 | 75 | |
GenBank mRNA | AB017156.1 | - | 8 - 9 | 1372 | 75 | |
AK008659.2 | - | 8 - 9 | 1386 | 75 | ||
AK033560.1 | - | 1390 | 75 | |||
AK136476.1 | - | 8 - 9 | 1386 | 75 | ||
BC028343.1 | - | 8 - 9 | 1379 | 4217548 | 75 | |
BC116318.2 | - | 8 - 9 | 1372 | 40095991 | 75 | |
BC116319.2 | - | 8 - 9 | 1372 | 40095992 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |