Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168568.1 | NP_001162040.1 | 17 - 18 | 1913 | 114 | |
NM_001168569.1 | NP_001162041.1 | 17 - 18 | 1970 | 114 | ||
NM_001168571.1 | NP_001162043.1 | 15 - 16 | 1892 | 114 | ||
NM_018737.5 | NP_061207.1 | 17 - 18 | 2045 | 114 | ||
NR_033143.1 | 17 - 18 | 1935 | 114 | |||
GenBank mRNA | AK032018.1 | - | 17 - 18 | 1772 | 114 | |
AK052854.1 | - | 17 - 18 | 1961 | 114 | ||
AK084595.1 | - | 17 - 18 | 1842 | 114 | ||
AK085381.1 | - | 17 - 18 | 1821 | 114 | ||
AK150966.1 | - | 17 - 18 | 1912 | 114 | ||
AK151597.1 | - | 17 - 18 | 1912 | 114 | ||
AK164206.1 | - | 15 - 16 | 1719 | 114 | ||
BC003257.1 | - | 17 - 18 | 1753 | 3500778 | 114 | |
EF410142.1 | - | 644 | 114 | |||
U49385.1 | - | 17 - 18 | 1935 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |