Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00020718.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020718.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020718.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020721.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020721.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020721.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020724.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020724.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020724.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020726.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020726.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020726.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020716.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020716.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020716.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020725.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020725.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020725.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020727.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020727.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020727.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020714.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020714.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020714.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020717.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020717.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020717.mm.1 |
TC0X00003327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020722.mm.1 |
NM_181315.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020722.mm.1 |
NM_181315.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020722.mm.1 |
TC0X00003327.mm.1 |
MTA TC |